Assay Details
Target Gene Details
Entrez Gene ID: | 70300 |
Gene Name: | fuzzy homolog (Drosophila) |
Gene Aliases: |
2600013E07Rik, b2b1273Clo |
Location: |
Chr.7:44895265-44900624 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fuz | NM_027376.3 | 1 | 141 | NP_081652.2 |
| AK011195.1 | 1 | 90 | BAB27459.1 | |
| AK033247.1 | 1 | 128 | ||
| AK134650.1 | 1 | 141 | BAE22226.1 | |
| AK168939.1 | 1 | 136 | ||
| BC045601.1 | 1 | 127 | AAH45601.1 |

Genomic Map