Assay Details
Target Gene Details
Entrez Gene ID: | 76858 |
Gene Name: | NLR family, pyrin domain containing 14 |
Gene Aliases: |
4921520L01Rik, GC-LRR, Nalp-iota, Nalp14, Nalp14l |
Location: |
Chr.7:107127432-107198103 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nlrp14 |
Target Gene Details
Entrez Gene ID: | 258597 |
Gene Name: | olfactory receptor 716 |
Gene Aliases: |
MOR260-2 |
Location: |
Chr.7:107147318-107148262 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr716 | NM_146604.1 | 1 | 471 | NP_666815.1 |
| BC120510.1 | 1 | 563 |
More Information
Set Membership: |
|

Genomic Map