Assay Details
Target Gene Details
Entrez Gene ID: | 269999 |
Gene Name: | ORAI calcium release-activated calcium modulator 3 |
Gene Aliases: |
9930124N15, BC061259, Tmem142c |
Location: |
Chr.7:127769589-127775150 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Orai3 | NM_198424.3 | 2 | 664 | NP_940816.1 |
| AB271216.1 | 2 | 490 | ||
| AK037138.1 | 3 | 808 | ||
| AK136955.1 | 2 | 665 | BAE23186.1 | |
| AK217284.1 | 1 | 53 | ||
| AM712358.1 | 2 | 490 | ||
| BC061259.1 | 2 | 634 | AAH61259.1 |
More Information
Set Membership: |
|

Genomic Map