Assay Details
Target Gene Details
Entrez Gene ID: | 13401 |
Gene Name: | dystrophia myotonica-containing WD repeat motif |
Gene Aliases: |
DMR-N9, Dm9 |
Location: |
Chr.7:19076089-19082776 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dmwd | NM_010058.2 | 5 | 2101 | NP_034188.2 |
| AK086899.1 | 4 | 1663 | ||
| BC031193.1 | 2 | 1195 | AAH31193.1 | |
| BC089027.1 | 4 | 2258 | AAH89027.1 | |
| S60312.1 | 5 | 1741 | AAC60663.1 |

Genomic Map