Assay Details
Target Gene Details
Entrez Gene ID: | 434198 |
Gene Name: | RIKEN cDNA B130024G19 gene |
Gene Aliases: |
- |
Location: |
Chr.7:70365383-70411146 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B130024G19Rik | NR_045850.1 | |||
| AK045070.1 | BAC32208.1 |
More Information
Set Membership: |
|

Genomic Map