Assay Details
Target Gene Details
Entrez Gene ID: | 381994 |
Gene Name: | RIKEN cDNA E030018B13 gene |
Gene Aliases: |
Gm1697 |
Location: |
Chr.7:63916857-63920539 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E030018B13Rik | NM_001256311.1 | 3 | 524 | NP_001243240.1 |
| AK086985.1 | 3 | 524 |
Target Gene Details
Entrez Gene ID: | 50794 |
Gene Name: | Kruppel-like factor 13 |
Gene Aliases: |
0610043C13Rik, 9430029L20Rik, Bteb3, FKLF-2, FKLF2, NSLP1, RFLAT-1, RFLAT1 |
Location: |
Chr.7:63886351-63938915 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klf13 | NM_021366.3 | NP_067341.2 | ||
| AF251796.1 | AAF73964.1 | |||
| AF252285.1 | AAF65826.1 | |||
| AJ245644.1 | CAB75887.1 | |||
| AJ275987.1 | CAC06697.1 | |||
| AK002926.1 | BAB22462.1 | |||
| AK155298.1 | BAE33174.1 |
More Information
Set Membership: |
|

Genomic Map