Assay Details
Target Gene Details
Entrez Gene ID: | 244233 |
Gene Name: | CD163 molecule-like 1 |
Gene Aliases: |
B430307C05, E430002D04Rik, Scart1 |
Location: |
Chr.7:140197218-140231145 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd163l1 |
Target Gene Details
Entrez Gene ID: | 258055 |
Gene Name: | olfactory receptor 524 |
Gene Aliases: |
MOR103-14P |
Location: |
Chr.7:140201336-140205279 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr524 | NM_001011814.1 | 1 | 801 | NP_001011814.1 |
| BC147558.1 | 1 | 833 | ||
| BC147561.1 | 1 | 833 |
More Information
Set Membership: |
|

Genomic Map