Assay Details
Target Gene Details
Entrez Gene ID: | 504186 |
Gene Name: | cholinergic receptor, nicotinic, alpha polypeptide 10 |
Gene Aliases: |
- |
Location: |
Chr.7:102111262-102116828 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrna10 | NM_001081424.1 | 5 | 1910 | NP_001074893.1 |
| AK033068.1 | 5 | 1910 | ||
| BC147245.1 | 5 | 1625 | ||
| BC147246.1 | 5 | 1625 | ||
| BC151033.1 | 5 | 1625 | ||
| BC151035.1 | 5 | 1625 |
More Information
Set Membership: |
|

Genomic Map