Assay Details
Target Gene Details
Entrez Gene ID: | 18631 |
Gene Name: | peroxisomal biogenesis factor 11 alpha |
Gene Aliases: |
PEX11alpha |
Location: |
Chr.7:79735957-79743126 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pex11a | NM_011068.1 | 3 | 667 | NP_035198.1 |
| AF093669.1 | 3 | 667 | AAC78659.1 | |
| AK076776.1 | 2 | 657 | ||
| AK078954.1 | 2 | 500 | BAC37477.1 | |
| AK149387.1 | 3 | 665 | BAE28847.1 | |
| BC023439.1 | 3 | 640 | AAH23439.1 |

Genomic Map