Assay Details
Target Gene Details
Entrez Gene ID: | 84113 |
Gene Name: | prostate tumor over expressed gene 1 |
Gene Aliases: |
1110030G05Rik, AU041779, Gcap3 |
Location: |
Chr.7:44863067-44870411 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptov1 | NM_133949.1 | 12 | 1827 | NP_598710.1 |
| AK003974.1 | BAB23101.1 | |||
| AK184856.1 | 1 | 67 | ||
| AK199978.1 | ||||
| AK200423.1 | ||||
| AK207880.1 | 2 | 108 | ||
| AK209436.1 | 2 | 108 | ||
| AK210324.1 | 2 | 108 | ||
| AK210403.1 | ||||
| AK213398.1 | 2 | 108 | ||
| AK217195.1 | 2 | 108 | ||
| BC008543.1 | 12 | 1703 | AAH08543.1 | |
| BC024632.1 | 12 | 1827 | AAH24632.1 |
More Information
Set Membership: |
|

Genomic Map