Assay Details
Target Gene Details
Entrez Gene ID: | 66367 |
Gene Name: | RIKEN cDNA 2310022A10 gene |
Gene Aliases: |
- |
Location: |
Chr.7:27553260-27582099 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2310022A10Rik | NM_001122767.1 | 9 | 1576 | NP_001116239.1 |
| NM_175107.5 | 10 | 2344 | NP_780316.3 | |
| AK009456.1 | 9 | 1546 | BAC25259.1 | |
| AK009941.1 | 9 | 1571 | ||
| BC023369.1 | 10 | 2323 | AAH23369.2 | |
| BQ560353.1 | 1 | 380 |

Genomic Map