Assay Details
Target Gene Details
Entrez Gene ID: | 11771 |
Gene Name: | adaptor-related protein complex 2, alpha 1 subunit |
Gene Aliases: |
Adtaa |
Location: |
Chr.7:44900373-44929490 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap2a1 | NM_001077264.1 | 23 | 3154 | NP_001070732.1 |
| NM_007458.2 | 24 | 3220 | NP_031484.1 | |
| AK135424.1 | 23 | 3083 | ||
| AK155515.1 | 23 | 3091 | ||
| BC031433.1 | 24 | 3170 | AAH31433.1 |
Target Gene Details
Entrez Gene ID: | 70300 |
Gene Name: | fuzzy homolog (Drosophila) |
Gene Aliases: |
2600013E07Rik, b2b1273Clo |
Location: |
Chr.7:44895265-44900624 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fuz | NM_027376.3 | NP_081652.2 | ||
| AK033247.1 | ||||
| AK134650.1 | BAE22226.1 | |||
| AK206194.1 | ||||
| BC045601.1 | AAH45601.1 | |||
| BC057127.1 | AAH57127.1 | |||
| CO041869.1 |
More Information
Set Membership: |
|

Genomic Map