Assay Details
Target Gene Details
Entrez Gene ID: | 386655 |
Gene Name: | EP300 interacting inhibitor of differentiation 2 |
Gene Aliases: |
Cri2, EID-2 |
Location: |
Chr.7:28267881-28269168 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eid2 | NM_198425.2 | 1 | 850 | NP_940817.1 |
| BC094513.1 | 1 | 850 | AAH94513.1 |

Genomic Map