Assay Details
Target Gene Details
Entrez Gene ID: | 67278 |
Gene Name: | PAXIP1 associated glutamate rich protein 1A |
Gene Aliases: |
2900092E17Rik, C77040, PA1, PAGR1 |
Location: |
Chr.7:127015051-127017352 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pagr1a | NM_030240.1 | 1 | 165 | NP_084516.1 |
| AK013840.1 | 1 | 142 | ||
| BC003932.1 | 1 | 165 | AAH03932.1 |

Genomic Map