Assay Details
Target Gene Details
Entrez Gene ID: | 11813 |
Gene Name: | apolipoprotein C-II |
Gene Aliases: |
- |
Location: |
Chr.7:19671579-19677941 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apoc2 | NM_001277944.1 | 5 | 597 | NP_001264873.1 |
| AA059961.1 | 2 | 243 | ||
| AK146468.1 | 4 | 398 | BAE27195.1 | |
| BC024697.1 | 5 | 597 | AAH24697.1 | |
| BG085960.2 | 4 | 440 | ||
| CB949564.1 | 4 | 503 | ||
| Z15090.1 | 4 | 398 | CAA78804.1 |
More Information
Set Membership: |
|

Genomic Map