Assay Details
Target Gene Details
Entrez Gene ID: | 73724 |
Gene Name: | methylmalonyl CoA epimerase |
Gene Aliases: |
1110007A04Rik |
Location: |
Chr.7:64392612-64412121 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mcee | NM_028626.1 | 2 | 129 | NP_082902.1 |
| AK003511.1 | 2 | 126 | BAB22828.1 | |
| BC038157.1 | 2 | 129 | AAH38157.1 |
More Information
Set Membership: |
|

Genomic Map