Assay Details
Target Gene Details
Entrez Gene ID: | 320871 |
Gene Name: | RIKEN cDNA B230206H07 gene |
Gene Aliases: |
- |
Location: |
Chr.7:141359178-141365110 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B230206H07Rik | NR_033532.1 | |||
| AK038776.1 | BAC30130.1 | |||
| AK080797.1 | 2 | 153 |
Target Gene Details
Entrez Gene ID: | 98845 |
Gene Name: | EPS8-like 2 |
Gene Aliases: |
AI042819, AW545405, Eps8r2 |
Location: |
Chr.7:141338911-141363016 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eps8l2 | NM_133191.2 | 21 | 2766 | NP_573454.2 |
| BC005492.1 | 21 | 2766 | AAH05492.1 | |
| BC009098.1 | 20 | 3050 | AAH09098.1 |
More Information
Set Membership: |
|

Genomic Map