Assay Details
Target Gene Details
Entrez Gene ID: | 101613 |
Gene Name: | NLR family, pyrin domain containing 6 |
Gene Aliases: |
AI504961, Avr, Nalp6, Navr, Navr/Avr, Pypaf5 |
Location: |
Chr.7:140920902-140929192 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nlrp6 | NM_133946.2 | 2 | 411 | NP_598707.2 |
FJ222825.1 | 2 | 360 | ||
JF810536.1 | 1 | 796 |
