Assay Details
Target Gene Details
Entrez Gene ID: | 69540 |
Gene Name: | kallikrein related-peptidase 10 |
Gene Aliases: |
2300002A13Rik, NES1, PRSSL1 |
Location: |
Chr.7:43781054-43785410 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klk10 | NM_133712.2 | 4 | 486 | NP_598473.1 |
| AK009000.1 | 4 | 471 | ||
| BC002100.1 | 4 | 485 | AAH02100.1 |
More Information
Set Membership: |
|

Genomic Map