Assay Details
Target Gene Details
Entrez Gene ID: | 11819 |
Gene Name: | nuclear receptor subfamily 2, group F, member 2 |
Gene Aliases: |
2700033K02Rik, 9430015G03Rik, ARP-1, Aporp1, COUP-TF2, COUP-TFII, COUPTFB, EAR3, SVP40, Tcfcoup2 |
Location: |
Chr.7:70351946-70366746 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nr2f2 | NM_009697.3 | 1 | 117 | NP_033827.2 |
| NM_183261.3 | NP_899084.2 | |||
| AK140125.1 | 1 | 41 | BAE24245.1 | |
| BC042484.1 | 1 | 117 | AAH42484.1 | |
| U07635.1 | AAA19854.1 |
More Information
Set Membership: |
|

Genomic Map