Assay Details
Target Gene Details
Entrez Gene ID: | 11819 |
Gene Name: | nuclear receptor subfamily 2, group F, member 2 |
Gene Aliases: |
2700033K02Rik, 9430015G03Rik, ARP-1, Aporp1, COUP-TF2, COUP-TFII, COUPTFB, EAR3, SVP40, Tcfcoup2 |
Location: |
Chr.7:70351946-70366746 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nr2f2 | NM_009697.3 | 3 | 3723 | NP_033827.2 |
| NM_183261.3 | 3 | 3671 | NP_899084.2 |
More Information
Set Membership: |
|

Genomic Map