Assay Details
Target Gene Details
Entrez Gene ID: | 17173 |
Gene Name: | achaete-scute complex homolog 2 (Drosophila) |
Gene Aliases: |
2410083I15Rik, Mash2, bHLHa45 |
Location: |
Chr.7:142966819-143009052 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ascl2 | NM_008554.3 | 2 | 428 | NP_032580.2 |
| AK010738.1 | 2 | 393 | ||
| AK167324.1 | 2 | 411 | BAE39426.1 | |
| BC019520.1 | 2 | 403 | AAH19520.1 | |
| CJ074389.1 |
More Information
Set Membership: |
|

Genomic Map