Assay Details
Target Gene Details
Entrez Gene ID: | 56787 |
Gene Name: | achaete-scute complex homolog 3 (Drosophila) |
Gene Aliases: |
Sgn1, bHLHa42 |
Location: |
Chr.7:109727465-109731732 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ascl3 | NM_020051.1 | 2 | 169 | NP_064435.1 |
| AB046448.1 | 2 | 155 | BAB83911.1 | |
| AB221543.1 | 1 | 109 | ||
| AJ277605.1 | 2 | 139 | CAC37689.1 | |
| BC104346.1 | 2 | 164 | AAI04347.1 | |
| BC104347.1 | 2 | 163 | AAI04348.1 |
More Information
Set Membership: |
|

Genomic Map