Assay Details
Target Gene Details
Entrez Gene ID: | 14132 |
Gene Name: | Fc receptor, IgG, alpha chain transporter |
Gene Aliases: |
FcRn |
Location: |
Chr.7:45092992-45103846 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fcgrt | NM_010189.3 | 2 | 284 | NP_034319.2 |
| AK008167.1 | 2 | 258 | ||
| AK008360.1 | 2 | 258 | ||
| AK146933.1 | 2 | 246 | ||
| AK152879.1 | 2 | 220 | BAE31564.1 | |
| BC003786.1 | 2 | 207 | AAH03786.1 | |
| BY063885.1 | ||||
| D37874.1 | 2 | 217 | BAA07111.1 |
More Information
Set Membership: |
|

Genomic Map