Assay Details
Target Gene Details
Entrez Gene ID: | 14132 |
Gene Name: | Fc receptor, IgG, alpha chain transporter |
Gene Aliases: |
FcRn |
Location: |
Chr.7:45092992-45103846 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fcgrt | NM_010189.3 | 3 | 631 | NP_034319.2 |
| AK146933.1 | 3 | 593 | ||
| AK152879.1 | 3 | 567 | BAE31564.1 | |
| BC003786.1 | 3 | 554 | AAH03786.1 | |
| D37874.1 | 3 | 564 | BAA07111.1 | |
| L17022.1 | 2 | 270 | AAA16904.1 |
More Information
Set Membership: |
|

Genomic Map