Assay Details
Target Gene Details
Entrez Gene ID: | 434128 |
Gene Name: | PNMA-like 2 |
Gene Aliases: |
EG434128 |
Location: |
Chr.7:16944682-16948828 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnmal2 | NM_001099636.2 | 1 | 1994 | NP_001093106.1 |

Genomic Map