Assay Details
Target Gene Details
Entrez Gene ID: | 66491 |
Gene Name: | polymerase (RNA) II (DNA directed) polypeptide L |
Gene Aliases: |
2510029B14Rik |
Location: |
Chr.7:141471860-141475153 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polr2l | NM_025593.1 | 2 | 1154 | NP_079869.1 |
| AK149137.1 | 2 | 1134 | BAE28747.1 |
More Information
Set Membership: |
|

Genomic Map