Assay Details
Target Gene Details
Entrez Gene ID: | 629378 |
Gene Name: | dapper homolog 3, antagonist of beta-catenin (xenopus) |
Gene Aliases: |
EG666671, naa-3 |
Location: |
Chr.7:16875317-16887301 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dact3 | NM_001081655.1 | 1 | 181 | NP_001075124.1 |
| AB073971.1 | 1 | 102 | ||
| DQ832319.1 | 1 | 181 |

Genomic Map