Assay Details
Target Gene Details
Entrez Gene ID: | 69757 |
Gene Name: | leukocyte receptor cluster (LRC) member 1 |
Gene Aliases: |
1500001K17Rik |
Location: |
Chr.7:3660107-3665840 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Leng1 | NM_027203.3 | 3 | 747 | NP_081479.1 |
| AK005095.1 | 3 | 378 | BAB23814.1 | |
| AK037634.1 | 3 | 394 | ||
| BC023108.1 | 3 | 747 | AAH23108.1 |

Genomic Map