Assay Details
Target Gene Details
Entrez Gene ID: | 321008 |
Gene Name: | RIKEN cDNA 6330408A02 gene |
Gene Aliases: |
- |
Location: |
Chr.7:13258967-13278721 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 6330408A02Rik | NM_177312.4 | 2 | 219 | NP_796286.2 |
| AK018141.1 | 2 | 182 | BAB31091.1 | |
| AK028865.1 | 2 | 168 | ||
| AK044247.1 | 1 | 1151 | ||
| AK132923.1 | 2 | 221 | BAE21424.1 | |
| AK160064.1 | 1 | 858 | BAE35600.1 | |
| AK162673.1 | 2 | 190 | BAE37016.1 | |
| BC075681.1 | 1 | 193 | AAH75681.1 | |
| CV783961.1 | 2 | 220 |
Target Gene Details
Entrez Gene ID: | 16881 |
Gene Name: | ligase I, DNA, ATP-dependent |
Gene Aliases: |
AL033288, LigI |
Location: |
Chr.7:13277313-13311427 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lig1 | NM_001199310.1 | NP_001186239.1 | ||
| BC138541.1 | ||||
| BC138542.1 | ||||
| U19604.1 | AAB60500.1 |
More Information
Set Membership: |
|

Genomic Map