Assay Details
Target Gene Details
Entrez Gene ID: | 23877 |
Gene Name: | Flt3 interacting zinc finger protein 1 |
Gene Aliases: |
AI790204, mFLJ00416 |
Location: |
Chr.7:5007056-5014728 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fiz1 | NM_001110328.1 | 2 | 447 | NP_001103798.1 |
| NM_001110329.1 | 2 | 390 | NP_001103799.1 | |
| NM_001110330.1 | 2 | 320 | NP_001103800.1 | |
| NM_011813.3 | 2 | 431 | NP_035943.3 | |
| AF126746.1 | 2 | 381 | AAD45498.1 | |
| AF126747.1 | 2 | 389 | AAD45499.1 | |
| AK135672.1 | 2 | 319 | ||
| AK139120.1 | 2 | 404 | ||
| AK142205.1 | 2 | 340 | BAE24975.1 | |
| AK162454.1 | 2 | 312 | BAE36926.1 | |
| AK170633.1 | 2 | 316 | BAE41925.1 | |
| AK196020.1 | 1 | 266 | ||
| AK220212.1 | 2 | 320 | BAD90137.1 | |
| BB855369.1 | ||||
| BC006633.1 | 2 | 431 | AAH06633.1 | |
| BC106113.1 | 2 | 306 | AAI06114.1 | |
| BC138231.1 | 2 | 381 | ||
| BC138232.1 | 2 | 406 | ||
| BC145207.1 | 2 | 406 | ||
| BU053482.1 |
More Information
Set Membership: |
|

Genomic Map