Assay Details
Target Gene Details
Entrez Gene ID: | 232791 |
Gene Name: | CCR4-NOT transcription complex, subunit 3 |
Gene Aliases: |
A930039N10Rik |
Location: |
Chr.7:3644522-3661109 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnot3 | NM_146176.3 | NP_666288.1 | ||
| AK141718.1 | ||||
| AK173003.1 | BAD32281.1 | |||
| BC030332.1 | AAH30332.1 | |||
| BC053437.1 | AAH53437.1 | |||
| BC141041.1 | ||||
| BQ748712.1 |
Target Gene Details
Entrez Gene ID: | 69757 |
Gene Name: | leukocyte receptor cluster (LRC) member 1 |
Gene Aliases: |
1500001K17Rik |
Location: |
Chr.7:3660107-3665840 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Leng1 | NM_027203.3 | 4 | 1587 | NP_081479.1 |
| AK037634.1 | 4 | 1235 | ||
| BC023108.1 | 4 | 1587 | AAH23108.1 |
More Information
Set Membership: |
|

Genomic Map