Assay Details
Target Gene Details
Entrez Gene ID: | 27397 |
Gene Name: | mitochondrial ribosomal protein L17 |
Gene Aliases: |
MRP-L26, Rpml26 |
Location: |
Chr.7:105803782-105811087 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl17 | NM_025301.2 | 1 | 57 | NP_079577.1 |
| AK007828.1 | BAB25288.1 | |||
| AK088838.1 | ||||
| AK144092.1 | BAE25695.1 | |||
| AK147957.1 | BAE28251.1 | |||
| BC010718.1 | AAH10718.1 | |||
| BY098205.1 | 1 | 69 |
More Information
Set Membership: |
|

Genomic Map