Assay Details
Target Gene Details
Entrez Gene ID: | 27397 |
Gene Name: | mitochondrial ribosomal protein L17 |
Gene Aliases: |
MRP-L26, Rpml26 |
Location: |
Chr.7:105803782-105811087 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl17 | NM_025301.2 | 3 | 3537 | NP_079577.1 |
| AK147957.1 | 3 | 3472 | BAE28251.1 |
More Information
Set Membership: |
|

Genomic Map