Assay Details
Target Gene Details
Entrez Gene ID: | 67315 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 12 |
Gene Aliases: |
1600031J20Rik, Ceacam12-C1, Ceacam12-C3, Ceacam12C1, Ceacam12C3 |
Location: |
Chr.7:18065929-18077986 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ceacam12 | NM_001162523.1 | 4 | 1012 | NP_001155995.1 |
| AK131656.1 | 4 | 1002 | BAE20744.1 |
More Information
Set Membership: |
|

Genomic Map