Assay Details
Target Gene Details
Entrez Gene ID: | 621080 |
Gene Name: | expressed sequence AI429214 |
Gene Aliases: |
- |
Location: |
Chr.8:36993575-36995533 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AI429214 | NM_001039220.3 | 1 | 332 | NP_001034309.1 |
| AK132736.1 | 1 | 304 | ||
| AK161334.1 | 1 | 326 | BAE36331.1 | |
| BC150875.1 | 1 | 285 | ||
| BY294724.1 | 1 | 333 |

Genomic Map