Assay Details
Target Gene Details
Entrez Gene ID: | 12564 |
Gene Name: | cadherin 8 |
Gene Aliases: |
AI851472, cad8 |
Location: |
Chr.8:99024471-99416485 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh8 | NM_001039154.2 | NP_001034243.1 | ||
| NM_001285913.1 | NP_001272842.1 | |||
| NM_001285914.1 | NP_001272843.1 | |||
| NM_007667.3 | 12 | 3060 | NP_031693.2 | |
| AK044046.1 | BAC31751.1 | |||
| AK083092.1 | BAC38758.1 | |||
| AK086711.1 | BAC39724.1 | |||
| AK139334.1 | BAE23961.1 | |||
| BC057581.1 | AAH57581.1 | |||
| X95600.1 | 12 | 2700 | CAA64857.1 |
Target Gene Details
Entrez Gene ID: | 100873092 |
Gene Name: | predicted gene 15679 |
Gene Aliases: |
OTTMUSG00000025218 |
Location: |
Chr.8:99007127-99032740 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm15679 | NR_110579.1 |
More Information
Set Membership: |
|

Genomic Map