Assay Details
Target Gene Details
Entrez Gene ID: | 70925 |
Gene Name: | CDKN2A interacting protein |
Gene Aliases: |
4921511I16Rik, AW208986, CARF |
Location: |
Chr.8:47709344-47714735 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdkn2aip | NM_172407.3 | 3 | 1037 | NP_765995.1 |
| AK014874.1 | 2 | 583 | ||
| AK048437.1 | 3 | 1006 | BAC33336.1 | |
| AK078329.1 | 3 | 1004 | ||
| AK089057.1 | 3 | 1007 | ||
| AK133831.1 | 3 | 1037 | ||
| AK167684.1 | 3 | 1005 | ||
| AK202731.1 | 1 | 183 |
More Information
Set Membership: |
|

Genomic Map