Assay Details
Target Gene Details
Entrez Gene ID: | 102060 |
Gene Name: | growth arrest and DNA-damage-inducible, gamma interacting protein 1 |
Gene Aliases: |
2310040G17Rik, AI425883, Crif1, MRP-L59, Plinp1 |
Location: |
Chr.8:84832282-84835482 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gadd45gip1 | NM_183358.4 | 2 | 1049 | NP_899202.3 |
| AK028227.1 | 2 | 1050 | BAC25826.1 |
Target Gene Details
Entrez Gene ID: | 19358 |
Gene Name: | RAD23a homolog (S. cerevisiae) |
Gene Aliases: |
2310040P19Rik, AL024030, HR23A, mHR23A |
Location: |
Chr.8:84834652-84840750 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rad23a | NM_001297606.1 | 9 | 1956 | NP_001284535.1 |
| NM_001297607.1 | 9 | 1891 | NP_001284536.1 | |
| NM_009010.5 | 9 | 1953 | NP_033036.2 | |
| AK038300.1 | 9 | 1930 | BAC29962.1 |
More Information
Set Membership: |
|

Genomic Map