Assay Details
Target Gene Details
Entrez Gene ID: | 11733 |
Gene Name: | ankyrin 1, erythroid |
Gene Aliases: |
Ank-1, nb, pale |
Location: |
Chr.8:22974836-23150502 on Build GRCm38 |
Assay Gene Location: | Within Exon 47 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ank1 | NM_001110783.3 | 44 | 6792 | NP_001104253.1 |
| NM_001277280.2 | 43 | 6303 | NP_001264209.1 | |
| NM_001277281.2 | 43 | 6489 | NP_001264210.1 | |
| NM_001277284.2 | 43 | 6564 | NP_001264213.1 | |
| NM_001277286.2 | 45 | 6700 | NP_001264215.1 | |
| NM_001277289.2 | 44 | 6778 | NP_001264218.1 | |
| NM_031158.4 | 45 | 6688 | NP_112435.2 | |
| NR_102385.2 | ||||
| NR_102386.2 | 45 | 6891 | ||
| AK134748.1 | 1 | 152 | ||
| AK147278.1 | 44 | 6747 | BAE27815.1 | |
| AK147483.1 | 45 | 6891 | ||
| AK147597.1 | 44 | 6726 | BAE28015.1 | |
| AK158131.1 | 43 | 6296 | BAE34375.1 | |
| BC079910.1 | 43 | 6535 | ||
| BC138029.1 | 45 | 6434 | ||
| BC171944.1 | 45 | 6434 | ||
| U73972.1 | 5 | 1483 | AAC24156.1 | |
| X69063.1 | 46 | 6672 | CAA48801.1 | |
| X69064.1 | 5 | 1447 | CAA48802.1 |
Target Gene Details
Entrez Gene ID: | 101056408 |
Gene Name: | predicted gene 15816 |
Gene Aliases: |
OTTMUSG00000026380 |
Location: |
Chr.8:23138785-23149585 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm15816 | NM_001282148.1 | NP_001269077.1 | ||
| AK042176.1 |
More Information
Set Membership: |
|

Genomic Map