Assay Details
Target Gene Details
Entrez Gene ID: | 15111 |
Gene Name: | heart and neural crest derivatives expressed transcript 2 |
Gene Aliases: |
AI225906, AI661148, Ehand2, Hed, Th2, Thing2, bHLHa26, dHAND |
Location: |
Chr.8:57320983-57324517 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hand2 | NM_010402.4 | 2 | 1982 | NP_034532.3 |
| AK035160.1 | 2 | 1984 | BAC28965.1 |
More Information
Set Membership: |
|

Genomic Map