Assay Details
Target Gene Details
Entrez Gene ID: | 18983 |
Gene Name: | CCR4-NOT transcription complex, subunit 7 |
Gene Aliases: |
AU022737, CAF-1, Caf1, Pop2 |
Location: |
Chr.8:40492538-40515847 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnot7 | NM_001271542.1 | NP_001258471.1 | ||
| AK135669.1 |
Target Gene Details
Entrez Gene ID: | 52348 |
Gene Name: | vacuolar protein sorting 37A (yeast) |
Gene Aliases: |
2210018P21Rik, 4930592A21Rik, AW261445, D8Ertd531e |
Location: |
Chr.8:40511783-40551134 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vps37a | NM_033560.3 | 1 | 302 | NP_291038.2 |
| AK016385.1 | 1 | 284 | ||
| AK048942.1 | 1 | 282 | ||
| AK054055.1 | 1 | 283 | ||
| AK137479.1 | 1 | 287 | BAE23372.1 | |
| AK157507.1 | 1 | 268 | ||
| AK170280.1 | 1 | 271 | ||
| AK170943.1 | 1 | 260 | BAE42131.1 | |
| AK170961.1 | 1 | 291 | BAE42142.1 | |
| AK171055.1 | 1 | 291 | BAE42215.1 | |
| AK171221.1 | 1 | 303 | BAE42324.1 | |
| BC039290.1 | 1 | 258 | AAH39290.1 | |
| BC110396.1 | 1 | 217 | ||
| BY085136.1 |
More Information
Set Membership: |
|

Genomic Map