Assay Details
Target Gene Details
Entrez Gene ID: | 56503 |
Gene Name: | ankyrin repeat domain 49 |
Gene Aliases: |
AW060934, C86108, Gbif |
Location: |
Chr.9:14779782-14784601 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd49 | NM_019683.3 | 1 | 177 | NP_062657.2 |
| AF028722.1 | 2 | 217 | AAD04809.1 | |
| AK132850.1 | 2 | 211 | BAE21393.1 | |
| AK137504.1 | 2 | 205 | ||
| AK153923.1 | 2 | 206 | BAE32257.1 | |
| AK155932.1 | 2 | 198 | ||
| AK157315.1 | 2 | 206 | ||
| BC019777.1 | 1 | 179 | AAH19777.1 |
More Information
Set Membership: |
|

Genomic Map