Assay Details
Target Gene Details
Entrez Gene ID: | 109006 |
Gene Name: | cytokine induced apoptosis inhibitor 1 |
Gene Aliases: |
2810413N20Rik, AA617265, AU021794, anamorsin |
Location: |
Chr.8:94819818-94838340 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ciapin1 | NM_134141.4 | 9 | 2633 | NP_598902.1 |
| AK043805.1 | 1 | 67 | ||
| AK136821.1 | 9 | 2627 | BAE23137.1 |

Genomic Map