Assay Details
Target Gene Details
Entrez Gene ID: | 67873 |
Gene Name: | methylthioribose-1-phosphate isomerase homolog (S. cerevisiae) |
Gene Aliases: |
2410018C20Rik |
Location: |
Chr.8:84249629-84257337 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mri1 | NM_026423.4 | 1 | 148 | NP_080699.3 |
| AK007709.1 | 1 | 110 | ||
| AK010538.1 | 1 | 120 | BAB27014.1 | |
| AK013882.1 | 1 | 117 | BAB29034.1 | |
| AK047287.1 | 1 | 151 | BAC33016.1 | |
| AK144807.1 | 1 | 88 | ||
| AK164635.1 | 1 | 148 | ||
| AK164894.1 | 1 | 117 | BAE37957.1 | |
| BC025049.1 | 1 | 115 | AAH25049.1 |
More Information
Set Membership: |
|

Genomic Map