Assay Details
Target Gene Details
Entrez Gene ID: | 69478 |
Gene Name: | RIKEN cDNA 2300009A05 gene |
Gene Aliases: |
- |
Location: |
Chr.9:63394447-63399244 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2300009A05Rik | NM_027090.1 | 1 | 182 | NP_081366.1 |
| AK009046.1 | 1 | 179 | BAB26045.1 | |
| AK054471.1 | 1 | 171 | ||
| AK158428.1 | 1 | 184 | BAE34504.1 | |
| BC116242.1 | 1 | 165 | ||
| BC116243.1 | 1 | 165 |

Genomic Map