Assay Details
Target Gene Details
Entrez Gene ID: | 75677 |
Gene Name: | claudin 22 |
Gene Aliases: |
2210404A22Rik, 5330431C14Rik, 9530051B05Rik |
Location: |
Chr.8:47824482-47825475 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cldn22 | NM_029383.1 | 1 | 819 | NP_083659.1 |
| AK008821.1 | 1 | 820 | BAB25914.1 |
More Information
Set Membership: |
|

Genomic Map