Assay Details
Target Gene Details
Entrez Gene ID: | 18983 |
Gene Name: | CCR4-NOT transcription complex, subunit 7 |
Gene Aliases: |
AU022737, CAF-1, Caf1, Pop2 |
Location: |
Chr.8:40492538-40515847 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnot7 | NM_001271542.1 | NP_001258471.1 | ||
| NM_001271543.1 | NP_001258472.1 | |||
| NM_011135.5 | NP_035265.1 | |||
| AK007767.1 | BAE43206.1 | |||
| AK009561.1 | ||||
| AK044535.1 | 1 | 219 | BAC31969.1 | |
| AK132063.1 | BAE20968.1 | |||
| AK135669.1 | ||||
| AK137838.1 | ||||
| AK147048.1 | BAE27634.1 | |||
| AK166449.1 | BAE38783.1 | |||
| BC006021.1 | AAH06021.1 | |||
| CJ166755.1 |
More Information
Set Membership: |
|

Genomic Map