Assay Details
Target Gene Details
Entrez Gene ID: | 79233 |
Gene Name: | zinc finger protein 319 |
Gene Aliases: |
AI225750, AI413472, Znf319, mKIAA1388 |
Location: |
Chr.8:95326136-95331950 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp319 | NM_024467.3 | 1 | 288 | NP_077787.3 |
| AK048624.1 | 1 | 148 | ||
| AK052274.1 | 1 | 123 | BAC34912.1 | |
| AK089263.1 | 1 | 138 | ||
| BC059823.1 | 1 | 288 | AAH59823.1 |
More Information
Set Membership: |
|

Genomic Map