Assay Details
Target Gene Details
Entrez Gene ID: | 73032 |
Gene Name: | tetratricopeptide repeat domain 9B |
Gene Aliases: |
2900074C18Rik |
Location: |
Chr.7:27653924-27656207 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc9b | NM_028417.1 | 3 | 708 | NP_082693.1 |
| AK013779.1 | 3 | 711 | BAB28991.1 | |
| BC089566.1 | 3 | 708 | AAH89566.1 |
More Information
Set Membership: |
|

Genomic Map